4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR27312-5GEA 0 DKK
APOSOR27312-5G APOSOR27312-1G
4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine
4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine
Formel: C₈H₈N₄S₂
Molekylvægt: 224,306 g/mol
Smeltepunkt: 176…177 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052772
CAS-nummer: 175204-61-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR