Ethyl 5-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR2689-250MGEA 1250 DKK
APOSOR2689-250MG
Ethyl 5-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥96%
Ethyl 5-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₁H₁₁ClN₂O₂
Molekylvægt: 238,67 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07366504
CAS-nummer: 141517-48-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR