3-Chloropropanamide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
APOSOR26786-5GEA
0
DKK
APOSOR26786-5G
APOSOR26786-1G
3-Chloropropanamide
3-Chloropropanamide
Formel:
C₃H₆ClNO Molekylvægt: 107,539 g/mol Smeltepunkt: 98…101 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008040 CAS-nummer: 5875-24-1 UN nummer: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...