3-Amino-1,2,3,4-tetrahydro-9H-carbazole-3-carboxylic acid, N9-BOC 3-FMOC protected 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR2676-100MGEA 1600 DKK
APOSOR2676-100MG APOSOR2676-250MG APOSOR2676-1G
3-Amino-1,2,3,4-tetrahydro-9H-carbazole-3-carboxylic acid, N9-BOC 3-FMOC protected 97%
3-Amino-1,2,3,4-tetrahydro-9H-carbazole-3-carboxylic acid, N9-BOC 3-FMOC protected
Formel: C₃₃H₃₂N₂O₆
Molekylvægt: 552,617 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09264213
CAS-nummer: 1031927-10-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR