2,2'-Bis(bromomethyl)-1,1'-biphenyl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
APOSOR26186-5GEA
0
DKK
APOSOR26186-5G
APOSOR26186-25G
2,2'-Bis(bromomethyl)-1,1'-biphenyl
2,2'-Bis(bromomethyl)-1,1'-biphenyl
Formel:
C₁₄H₁₂Br₂ Molekylvægt: 340,053 g/mol Smeltepunkt: 90…92 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00074910 CAS-nummer: 38274-14-5 UN nummer: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...