2,3-Dihydro-1,4-benzodioxine-5-carboxamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR25997-1GEA 0 DKK
APOSOR25997-1G APOSOR25997-5G APOSOR25997-100MG
2,3-Dihydro-1,4-benzodioxine-5-carboxamide 97%
2,3-Dihydro-1,4-benzodioxine-5-carboxamide
Formel: C₉H₉NO₃
Molekylvægt: 179,173 g/mol
Smeltepunkt: 130…132 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03659722
CAS-nummer: 349550-81-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR