3,5-Dibromo-2-ethoxybenzaldehyde
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR25780-1GEA
590
DKK
APOSOR25780-1G
3,5-Dibromo-2-ethoxybenzaldehyde
3,5-Dibromo-2-ethoxybenzaldehyde
Formel:
C₉H₈Br₂O₂ Molekylvægt: 307,967 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00087286 CAS-nummer: 61657-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...