1-Isopropyluracil-5-carbonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR25579-1GEA 1100 DKK
APOSOR25579-1G
1-Isopropyluracil-5-carbonitrile
1-Isopropyluracil-5-carbonitrile
Formel: C₈H₉N₃O₂
Molekylvægt: 179,176 g/mol
Smeltepunkt: 265…274 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00203975
CAS-nummer: 57712-59-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR