Cyclohexyl chloroacetate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

APOSOR25297-10GEA 0 DKK
APOSOR25297-10G APOSOR25297-1G
Cyclohexyl chloroacetate
Cyclohexyl chloroacetate
Formel: C₈H₁₃ClO₂
Molekylvægt: 176,641 g/mol
Kogepunkt: 102…105 °C (15 mm Hg)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00084989
CAS-nummer: 6975-91-3
UN nummer: 3265
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR