Ethyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR25286-1GEA 1160 DKK
APOSOR25286-1G
Ethyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate
Ethyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate
Formel: C₉H₁₀N₂O₂S₂
Molekylvægt: 242,318 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00085050
CAS-nummer: 116170-90-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR