Ethyl (4,5-dichloro-1H-imidazol-1-yl)acetate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

|
Advarsel
|
APOSOR25100-100MGEA
273
DKK
APOSOR25100-100MG
Ethyl (4,5-dichloro-1H-imidazol-1-yl)acetate
Ethyl (4,5-dichloro-1H-imidazol-1-yl)acetate
Formel:
C₇H₈Cl₂N₂O₂ Molekylvægt: 223,057 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00052518 CAS-nummer: 175137-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...