1-Methyl-5-(pyridin-2-yl)pyrrolidin-2-one

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR2505-10MGEA 0 DKK
APOSOR2505-10MG APOSOR2505-5MG
1-Methyl-5-(pyridin-2-yl)pyrrolidin-2-one
1-Methyl-5-(pyridin-2-yl)pyrrolidin-2-one

A metabolite of nicotine in humans. Used for internal standardization for routine biomonitoring of nicotine metabolites.

Formel: C₁₀H₁₂N₂O
Molekylvægt: 176,215 g/mol
Kogepunkt: 120…135 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01320387
CAS-nummer: 147732-31-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR