(2,4-Dichlorophenoxy)acetyl chloride
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
APOSOR2440-100MGEA
176
DKK
APOSOR2440-100MG
APOSOR2440-250MG
(2,4-Dichlorophenoxy)acetyl chloride
(2,4-Dichlorophenoxy)acetyl chloride
Formel:
C₈H₅Cl₃O₂ Molekylvægt: 239,483 g/mol Kogepunkt: 182 °C (15 mm Hg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00045225 CAS-nummer: 774-74-3 UN nummer: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...