1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR24326-1GEA 933 DKK
APOSOR24326-1G
1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid
1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₃H₁₄N₂O₂
Molekylvægt: 230,262 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00068140
CAS-nummer: 116344-17-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR