Ethyl 5-propyl-1H-pyrazole-4-carboxylate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR24284-500MGEA
154
DKK
APOSOR24284-500MG
Ethyl 5-propyl-1H-pyrazole-4-carboxylate
Ethyl 5-propyl-1H-pyrazole-4-carboxylate
Formel:
C₉H₁₄N₂O₂ Molekylvægt: 182,22 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01570118 CAS-nummer: 123374-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...