2-(2-Nitrophenyl)acrylaldehyde
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

APOSOR23876-1GEA
579
DKK
APOSOR23876-1G
APOSOR23876-10G
2-(2-Nitrophenyl)acrylaldehyde
2-(2-Nitrophenyl)acrylaldehyde
Formel:
C₉H₇NO₃ Molekylvægt: 177,157 g/mol Smeltepunkt: 53…55 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00052673 CAS-nummer: 71463-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...