3-Amino-3,4-dihydro-4-oxo-N-[(tetrahydrofuran-2-yl)methyl]quinazoline-2-carboxamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR23565-250MGEA 197 DKK
APOSOR23565-250MG
3-Amino-3,4-dihydro-4-oxo-N-[(tetrahydrofuran-2-yl)methyl]quinazoline-2-carboxamide
3-Amino-3,4-dihydro-4-oxo-N-[(tetrahydrofuran-2-yl)methyl]quinazoline-2-carboxamide
Formel: C₁₄H₁₆N₄O₃
Molekylvægt: 288,302 g/mol
Smeltepunkt: 154…156 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01566065
CAS-nummer: 646497-34-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR