[3-(1H-Pyrrol-1-yl)phenyl]methanol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR23351-1GEA
634
DKK
APOSOR23351-1G
[3-(1H-Pyrrol-1-yl)phenyl]methanol
[3-(1H-Pyrrol-1-yl)phenyl]methanol
Formel:
C₁₁H₁₁NO Molekylvægt: 173,211 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03086142 CAS-nummer: 83140-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...