4-(4-Bromophenyl)-1,2,3-thiadiazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR23263-1GEA 459 DKK
APOSOR23263-1G APOSOR23263-10G
4-(4-Bromophenyl)-1,2,3-thiadiazole
4-(4-Bromophenyl)-1,2,3-thiadiazole
Formel: C₈H₅BrN₂S
Molekylvægt: 241,108 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00084907
CAS-nummer: 40753-13-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR