4-Iodo-5-methyl-1-phenyl-1H-pyrazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

|
Advarsel
|
APOSOR23250-250MGEA
197
DKK
APOSOR23250-250MG
4-Iodo-5-methyl-1-phenyl-1H-pyrazole
4-Iodo-5-methyl-1-phenyl-1H-pyrazole
Formel:
C₁₀H₉IN₂ Molekylvægt: 284,096 g/mol Smeltepunkt: 65…68 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02681924 CAS-nummer: 342405-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...