1-Benzofuran-5-carbonyl chloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

APOSOR23099-1GEA 0 DKK
APOSOR23099-1G
1-Benzofuran-5-carbonyl chloride
1-Benzofuran-5-carbonyl chloride
Formel: C₉H₅ClO₂
Molekylvægt: 180,588 g/mol
Smeltepunkt: 67…69 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03659699
CAS-nummer: 56540-70-6
UN nummer: 3261
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR