3,5-Dimethylisoxazol-4-yl isothiocyanate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR22988-250MGEA 0 DKK
APOSOR22988-250MG APOSOR22988-1G
3,5-Dimethylisoxazol-4-yl isothiocyanate
3,5-Dimethylisoxazol-4-yl isothiocyanate
Formel: C₆H₆N₂OS
Molekylvægt: 154,19 g/mol
Kogepunkt: 62…63 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02681978
CAS-nummer: 321309-27-7
UN nummer: 2810
ADR: 6.1,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR