2-Amino-5-(hydroxymethyl)pyrazine, 2-BOC protected

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR2290-50MGEA 383 DKK
APOSOR2290-50MG
2-Amino-5-(hydroxymethyl)pyrazine, 2-BOC protected
2-Amino-5-(hydroxymethyl)pyrazine, 2-BOC protected
Formel: C₁₀H₁₅N₃O₃
Molekylvægt: 225,244 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09952117
CAS-nummer: 874476-55-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR