2-Methyl-5-sulphamoylbenzoic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR2277-1GEA
656
DKK
APOSOR2277-1G
2-Methyl-5-sulphamoylbenzoic acid
2-Methyl-5-sulphamoylbenzoic acid
Formel:
C₈H₉NO₄S Molekylvægt: 215,226 g/mol Smeltepunkt: 243…250 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07362354 CAS-nummer: 20532-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...