N-[2-(2-Amino-1,3-thiazol-5-yl)ethyl]phthalimide hydrobromide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR2274-1GEA 0 DKK
APOSOR2274-1G APOSOR2274-5G
N-[2-(2-Amino-1,3-thiazol-5-yl)ethyl]phthalimide hydrobromide
N-[2-(2-Amino-1,3-thiazol-5-yl)ethyl]phthalimide hydrobromide
Formel: C₁₃H₁₁N₃O₂S·BrH
Molekylvægt: 354,222 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09836180
CAS-nummer: 136604-50-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR