1-Aminocyclooctanecarboxylic acid, N-FMOC protected 99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR2233-1GEA 4150 DKK
APOSOR2233-1G
1-Aminocyclooctanecarboxylic acid, N-FMOC protected 99%
1-Aminocyclooctanecarboxylic acid, N-FMOC protected
Formel: C₂₄H₂₇NO₄
Molekylvægt: 393,475 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03426230
CAS-nummer: 222166-38-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR