2-Phenoxypyridine-3-carbonyl chloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Farlig

APOSOR21221-1GEA 415 DKK
APOSOR21221-1G APOSOR21221-5G
2-Phenoxypyridine-3-carbonyl chloride
2-Phenoxypyridine-3-carbonyl chloride
Formel: C₁₂H₈ClNO₂
Molekylvægt: 233,65 g/mol
Smeltepunkt: 94…95 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052648
CAS-nummer: 51362-49-3
UN nummer: 1759
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR