Methyl 5-(naphth-2-yl)-1H-pyrazole-3-carboxylate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR1987-500MGEA 0 DKK
APOSOR1987-500MG APOSOR1987-1G
Methyl 5-(naphth-2-yl)-1H-pyrazole-3-carboxylate
Methyl 5-(naphth-2-yl)-1H-pyrazole-3-carboxylate
Formel: C₁₅H₁₂N₂O₂
Molekylvægt: 252,268 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06245485
CAS-nummer: 164295-93-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR