Diethyl (1-ethoxyethylidene)malonate 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR18414-1GEA
0
DKK
APOSOR18414-1G
APOSOR18414-250MG
Diethyl (1-ethoxyethylidene)malonate 97%
Diethyl (1-ethoxyethylidene)malonate
Formel:
C₁₁H₁₈O₅ Molekylvægt: 230,258 g/mol Kogepunkt: 123…128 °C (3 mm Hg) Smeltepunkt: 25…27 °C Massefylde: 1,068 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD22575006 CAS-nummer: 3044-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...