[1-(3-Amino-4-nitrophenyl)piperidin-4-yl]methanol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR183522-1GEA 1370 DKK
APOSOR183522-1G
[1-(3-Amino-4-nitrophenyl)piperidin-4-yl]methanol
[1-(3-Amino-4-nitrophenyl)piperidin-4-yl]methanol
Formel: C₁₂H₁₇N₃O₃
Molekylvægt: 251,282 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 1604046-43-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR