6-Bromo-3-(dimethylamino)-1,2,3,4-tetrahydro-9H-carbazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR17505-1GEA
546
DKK
APOSOR17505-1G
APOSOR17505-5G
6-Bromo-3-(dimethylamino)-1,2,3,4-tetrahydro-9H-carbazole
6-Bromo-3-(dimethylamino)-1,2,3,4-tetrahydro-9H-carbazole
Useful as 5-HT1F receptor antagonists
Formel:
C₁₄H₁₇BrN₂ Molekylvægt: 293,202 g/mol Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD13563140 CAS-nummer: 186545-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...