1-Ethyl-3-methyl-1H-pyrazole-5-carbonyl chloride
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

|
Farlig
|
APOSOR1519-1GEA
1100
DKK
APOSOR1519-1G
1-Ethyl-3-methyl-1H-pyrazole-5-carbonyl chloride
1-Ethyl-3-methyl-1H-pyrazole-5-carbonyl chloride
Formel:
C₇H₉ClN₂O Molekylvægt: 172,612 g/mol Kogepunkt: 112 °C (15 mm Hg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01313846 CAS-nummer: 128249-59-2 UN nummer: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...