4,4,6-Trimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR14179-1GEA 0 DKK
APOSOR14179-1G APOSOR14179-5G
4,4,6-Trimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
4,4,6-Trimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
Formel: C₁₃H₁₉BO₂
Molekylvægt: 218,1 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08436973
CAS-nummer: 1310404-79-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR