2-(Aminomethyl)pyrimidine oxalate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR12993-50MGEA 743 DKK
APOSOR12993-50MG
2-(Aminomethyl)pyrimidine oxalate
2-(Aminomethyl)pyrimidine oxalate
Formel: C₅H₇N₃·C₂H₂O₄
Molekylvægt: 199,164 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04117723
CAS-nummer: 866625-10-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR