3-Benzylmorpholine hydrochloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR12969-250MGEA 0 DKK
APOSOR12969-250MG APOSOR12969-1G APOSOR12969-100MG
3-Benzylmorpholine hydrochloride
3-Benzylmorpholine hydrochloride
Formel: C₁₁H₁₅NO·ClH
Molekylvægt: 213,704 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04117744
CAS-nummer: 1172897-29-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR