3-(4-Methoxyphenyl)morpholine hydrochloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR12851-500MGEA 0 DKK
APOSOR12851-500MG APOSOR12851-100MG
3-(4-Methoxyphenyl)morpholine hydrochloride
3-(4-Methoxyphenyl)morpholine hydrochloride
Formel: C₁₁H₁₅NO₂·ClH
Molekylvægt: 229,703 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03840088
CAS-nummer: 1171560-33-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR