4-(2-Amino-1-phenylethyl)piperazine, N1-BOC protected

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR12704-1GEA 0 DKK
APOSOR12704-1G
4-(2-Amino-1-phenylethyl)piperazine, N1-BOC protected
4-(2-Amino-1-phenylethyl)piperazine, N1-BOC protected
Formel: C₁₇H₂₇N₃O₂
Molekylvægt: 305,415 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01862526
CAS-nummer: 444892-54-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR