Quantitative filter papers, cone folded, grade 40
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Whatman™
WHAT990010116EA 6190 DKK
WHAT990010116 WHAT990010112
Quantitative filter papers, cone folded, grade 40
Filtre Filtrerpapir Kvantitativt filtrerpapir
Whatman™ cellulose filter paper grades are available in ready-to-use cone folded formats.
  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency that you expect from all Whatman™ filter papers
Maintains flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself. There are various stacking and packaging options available to suit customer needs.