Advanced hand sanitiser, green certified foam, PURELL®
Leverandør: Gojo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PURELL®
GOJO1904-02-EEU00EA 208 DKK
GOJO1904-02-EEU00
Advanced hand sanitiser, green certified foam, PURELL®
Hudpleje Cremer Hand and Body Lotions
Luxurious foam made with naturally-renewable ethanol. Kills 99,99 percent of most common germs that may cause illness.
  • Removable pump for easier recycling
  • SANITARY SEALED™ refill helps prevent contamination
  • Clinically-proven to maintain skin health
Patent-pending controlled collapse technology maintains bottle shape longer while emptying. SMART-FLEX™ bottle design uses 30 percent less material than HDPE.
Dispenser material is durable ABS Plastic with rugged polycarbonate view windows (both recycling code number 7).
Certificeringer: ECOLOGO® Certified to Hand Sanitizer Standard UL 2783. USDA Certified Biobased Product.
Advarsel: Use biocides safely. Always read the label and product information before use.