VWR®, Sealing films, Non Tacky, for qPCR, Storage and Crystallisation
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
391-0644EA 625 DKK
391-0644 391-0624 391-0189
VWR®, Sealing films, Non Tacky, for qPCR, Storage and Crystallisation
Forsegling Mikrotiterpladetætninger
50 µm polyolefin films with 50 µm inert encapsulated silicone adhesive. Especially suited for qPCR, storage and protein crystallisation applications. The encapsulated silicone adhesive is non tacky until pressed against the sealing surface, at which time adhesive is released only in sealing areas to form the strongest available heat resistant seal around each well on the plate.
  • High optical clarity, minimal to no autofluorescence
  • Chemically inert; no extractables except at extreme pH; DMSO resistant for HTS
  • Heat resistant, recommended for temperatures from −80 to +105 °C
  • Certified free from DNase, RNase, and nucleic acid
Sized to fit within the edges of raised rim 96-well plates (76,2×133,4 mm). Two end tabs assist in positioning the film on the plate.