Fan attachment, 1040 series, standard, Cool Flow™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Cool Flow™
MMMA1040EA 1960 DKK
MMMA1040
Fan attachment, 1040 series, standard, Cool Flow™
Åndedrætsværn Genanvendelige masker Halvmasker
The Cool Flow™ Fan 1040 is an optional attachment for 3M 4000+ Series Reusable Half Masks. This fan features a single button control function, offering improved wearer comfort during long wok shifts.
  • Reduces heat and moisture build up
  • Optional attachment that attaches simply and securely
  • Noticeable cooling sensation on the face
  • Single button control function
  • Rechargeable and supplied with a European plug
Reduce heat and moisture build up inside a half-face respirator with the Cool Flow™ Fan 1040. This innovative attachment for 3M 4000+ Series Reusable Half Masks keeps workers cool and dry over long work shifts. The fan provides a noticeable cooling sensation on the face, and provides additional comfort without compromising breathing protection. It attaches simply and securely to the front of the mask and features a single button control function to adjust airflow. In addition to hot environments, it’s ideal for a range of workplaces and activities. It is rechargeable and is supplied with a European plug.