Clofenotan
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1,1,1-Trichlor-2,2-bis(4-chlorphenyl)ethan
, Dichlordiphenyltrichlorethan
, DDT
, Dicophan
EHERC12080000EA
989
DKK
EHERC12080000
Clofenotan
Clofenotan
Formel:
C₁₄H₉Cl₅ Molekylvægt: 354,49 g/mol Kogepunkt: 260 °C (1013 hPa) Smeltepunkt: ∼89 °C Massefylde: ∼1,5 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
CAS-nummer:
50-29-3 EINECS: 200-024-3 UN nummer: 2761 ADR: 6.1,III Merck Index: 12,02898 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...