Tolbutamide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAT3690-1GEA 132 DKK
TCIAT3690-1G
Tolbutamide ≥98.0% (by HPLC, titration analysis)
Tolbutamide
Formel: C₁₂H₁₈N₂O₃S
MDL Number: MFCD00027169
CAS-nummer: 64-77-7
EINECS: 200-594-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 130.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR