2,3,3,5-Tetramethylindolenine ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAT3458-25GEA 1360 DKK
TCIAT3458-25G TCIAT3458-5G
2,3,3,5-Tetramethylindolenine ≥98.0% (by GC)
2,3,3,5-Tetramethylindolenine
Formel: C₁₂H₁₅N
Kogepunkt: 125°C
MDL Number: MFCD03093020
CAS-nummer: 25981-82-2

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR