Tricyclohexylphosphine oxide ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAT3430-1GEA 1250 DKK
TCIAT3430-1G
Tricyclohexylphosphine oxide ≥98.0% (by GC)
Tricyclohexylphosphine oxide
Formel: C₁₈H₃₃OP
Molekylvægt: 296,42 g/mol
Kogepunkt: 210°C
MDL Number: MFCD00014298
CAS-nummer: 13689-19-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 155.0 to 159.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR