Theophylline-7-acetic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAT2941-25GEA
206
DKK
TCIAT2941-25G
Theophylline-7-acetic acid ≥98.0% (by HPLC, titration analysis)
Theophylline-7-acetic acid
Formel:
C₉H₁₀N₄O₄ Molekylvægt: 238,20 |
MDL Number:
MFCD00022832 CAS-nummer: 652-37-9 EINECS: 211-490-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...