Thiophene-3-methylamine ≥97.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAT2850-1GEA
772
DKK
TCIAT2850-1G
Thiophene-3-methylamine ≥97.0% (by GC, titration analysis)
Thiophene-3-methylamine
Formel:
C₅H₇NS Molekylvægt: 113,18 g/mol |
MDL Number:
MFCD01529872 CAS-nummer: 27757-86-4 UN nummer: 2735 ADR: 8,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...