4-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAT2841-1GEA
197
DKK
TCIAT2841-1G
4-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Carboxyphenylboronic acid pinacol ester
Formel:
C₁₃H₁₇BO₄ Molekylvægt: 248,08 g/mol |
MDL Number:
MFCD01863710 CAS-nummer: 180516-87-4 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 228.0 to 232.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...