Pyren ≥98.0% (by GC), purified by sublimation

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAP2072-1GEA 288 DKK
TCIAP2072-1G
Pyren ≥98.0% (by GC), purified by sublimation
Pyrene
Formel: C₁₆H₁₀
Molekylvægt: 202,26 g/mol
Kogepunkt: 404°C
MDL Number: MFCD00004136
CAS-nummer: 129-00-0
EINECS: 204-927-3
UN nummer: 2811
ADR: 6.1,II
https://dk.vwr.com/stibo/web/std.lang.all/32/92/28993292.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR