(2-Nitrophenyl)hydrazine hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAN0231-25GEA
1790
DKK
TCIAN0231-25G
(2-Nitrophenyl)hydrazine hydrochloride ≥98.0% (by titrimetric analysis)
(2-Nitrophenyl)hydrazine hydrochloride
Formel:
C₆H₈ClN₃O₂ Molekylvægt: 189,6 g/mol |
MDL Number:
MFCD00060205 CAS-nummer: 6293-87-4 UN nummer: 1325 ADR: 4.1,II |
Specifikationer Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (Potassium iodate Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...